Learn More
CD42b/GPIb alpha Antibody (MM2/174), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128501
Catalog No. NBP128501
Only null left
Add to Cart
Description
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD42b/GPIb alpha
Monoclonal
FITC
Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit
Mouse
Protein A or G purified
RUO
2811
Human
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
MM2/174
Flow Cytometry 10 μL/10^6 cells
GP1BA
Human plasma membrane
1 mL
Primary
Human CD42b
Store at 4C. Do not freeze.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only