Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD42b/GPIb alpha Antibody (SR1163), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321781100UL
Description
CD42b/GPIb alpha Monoclonal antibody specifically detects CD42b/GPIb alpha in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
CD42b/GPIb alpha | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1163 | |
Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200 | |
Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
A synthesized peptide derived from human CD42b/GPIb alpha (Uniprot #: P07359) | |
100 μg | |
Immunology | |
2811 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction