Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD42b Monoclonal Antibody (HIP1), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD42b |
---|---|
Clone | HIP1 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H051T03B08A
![]() |
Invitrogen™
H051T03B08A |
100 Tests |
Each for $430.00
|
|
|||||
H051T02B08A
![]() |
Invitrogen™
H051T02B08A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD42b Monoclonal antibody specifically detects CD42b in Human samples. It is validated for Flow CytometrySpecifications
CD42b | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P07359 | |
2811 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HIP1 | |
Flow Cytometry | |
NovaFluor Blue 660-120S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP | |
IgG1 κ | |
Affinity chromatography | |
Gp1ba |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title