Learn More
CD42d Mouse Anti-Rat, Brilliant Violet 650, Clone: RPM.4, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743620
Description
The RPM.4 monoclonal antibody specifically recognizes CD42d (Platelet glycoprotein V, GPV). CD42d is a 88-kDa type I transmembrane glycoprotein which noncovalently associates with GPIb and GPIX to form a receptor for von Willebrand factor on megakaryocytes and resting platelets. The binding sites for von Willebrand factor and thrombin have been localized to the GPIb chain of the GPI-b-V-IX complex. GPV is cleaved by thrombin to release a 70 kDa soluble fragment, which contains the epitope recognized by mAb RPM.4. Although the rat CD42d protein's structure is similar to that of human CD42d, the RPM.4 antibody does not cross-react with human platelets.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
CD42d | |
RPM.4 | |
Brilliant Violet 650 | |
PLGPV, Gp5, platelet glycoprotein V, GPV, Platelet glycoprotein 5 | |
Wistar rat platelets | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.