Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD42d Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119035
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119035 200 μL
PIPA5122111 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119035 Supplier Invitrogen™ Supplier No. PA5119035
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD42d Polyclonal Antibody for Western Blot, IHC (P), Flow

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42d
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Gp5
Gene Accession No. P40197
Gene Alias CD42d; Glycoprotein 5; glycoprotein 5 (platelet); glycoprotein 5, platelet; glycoprotein V (platelet); glycoprotein V platelet; GP5; GPV; Platelet glycoprotein 5; platelet glycoprotein V; Platelete glycoprotein 5; Platelets GP V; PLGPV
Gene Symbols Gp5
Host Species Rabbit
Immunogen Recombinant protein Cluster of Differentiation 42d. The antigen corresponds to amino acid range 17-242 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.