Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD43 (Leukosialin), Rat Anti-Mouse, RB613, Clone: S7, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 759352
Description
CD43 (Leukosialin) Monoclonal antibody specifically detects CD43 (Leukosialin) in Mouse (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
CD43 (Leukosialin) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
20737 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Flow Cytometry | |
S7 | |
RB613 | |
Spn; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK | |
Mouse Plasmacytoma MOPC-315 | |
50 μg; | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction