Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD43 Monoclonal Antibody (eBio84-3C1 (84-3C1)), PE-Cyanine7, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 25043942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-043-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-043-942 Supplier Invitrogen™ Supplier No. 25043942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAb ebio84-3C1 reacts with CD43, which is also known as leukosialin, galactoglycoprotein and sialophorin. CD43 is a sialomucin which like many mucins can have both adhesive and anti-adhesive functions. Expression of CD43 is found on most leukocytes except resting B lymphocytes. Proteolytic processing upon activation decreases surface expression. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes. The counter-receptor for CD43 is CD169/SIGLEC-1, which is expressed on macrophages.

CD43, also known as leukosialin or sialophorin, is a transmembrane mucin-like protein characterized by a high negative charge. It is expressed on the surface of most hematopoietic cells, excluding resting B lymphocytes. CD43 serves both adhesive and anti-adhesive functions, contributing to a repulsive barrier that can interfere with cellular adhesion while also promoting leukocyte aggregation in certain contexts. CD43 is involved in the activation of T cells, B cells, NK cells, and monocytes. Its expression decreases upon activation due to proteolytic processing. The counter-receptor for CD43 is CD169/SIGLEC-1, which is expressed on macrophages. CD43 interacts with actin-binding proteins ezrin and moesin, playing a regulatory role in remodeling T-cell morphology and regulating cell-cell interactions during lymphocyte traffic. It enhances LFA-1 adhesiveness while counteracting LFA-1 induction via other receptors. CD43 signaling induces the functionally active tumor suppressor p53 protein, but in cases of p53 and ARF deficiency, it can promote tumor proliferation and viability. CD43 is an important modulator of leukocyte functions, with dysfunctions associated with diseases such as Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD43
Applications Flow Cytometry
Classification Monoclonal
Clone eBio84-3C1 (84-3C1)
Concentration 5 μL/Test
Conjugate PE-Cyanine7
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene SPN
Gene Accession No. P16150
Gene Alias 3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen
Gene Symbols SPN
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6693
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.