Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43 Mouse, anti-Human, R718, Clone: L60, BD OptiBuild™

Catalog No. BDB752087 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752087 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752087 Supplier BD Biosciences Supplier No. 752087
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L60 monoclonal antibody specifically binds to CD43 which is also known as Leukosialin (LSN) or Galactoglycoprotein (GALGP). CD43 is a ∼95-135 kDa heavily O-sialylated type I transmembrane glycoprotein that is encoded by SPN (sialophorin) and belongs to the cell surface mucin family. The L60 antibody recognizes a sialic acid-dependent determinant on CD43. CD43 is highly expressed on T lymphocytes, thymocytes, monocytes, granulocytes, bone marrow stem cells, pre-B cells and activated B cells plasma cells but not on resting peripheral blood B cells, red blood cells, and non-hematopoietic cells. CD43 is enzymatically shed from leucocyte surfaces following activation by various stimuli. CD43 appears to be involved in mediating intercellular interactions that regulate leucocyte functions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD43
Applications Flow Cytometry
Classification Monoclonal
Clone L60
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.