Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD43/Sialophorin Antibody (84-3C1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311445AF532
Description
CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CD43/Sialophorin | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
84-3C1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) | |
Stimulated human leukocytes | |
0.1 mL | |
B Cell Development and Differentiation Markers, Immunology | |
6693 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction