Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD43/Sialophorin Antibody (MEM-59), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262227MFV450
Description
CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Human samples. It is validated for Flow CytometrySpecifications
CD43/Sialophorin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MEM-59 | |
Flow Cytometry | |
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) | |
Human T lymphocytes. | |
0.1 mL | |
B Cell Development and Differentiation Markers, Immunology | |
6693 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction