Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Antibody (CD44v4/1219) - variant 4, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00 - $536.00

Specifications

Antigen CD44, variant 4
Clone CD44v4/1219
Concentration 0.2 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25319720
SDP
View Documents
Novus Biologicals
NBP25319720UG
20 μg
Each for $280.00
Only null left
Add to Cart
 
NB253197100
SDP
View Documents
Novus Biologicals
NBP253197100UG
100 μg
Each for $536.00
Only null left
Add to Cart
 
NB253197200
SDP
View Documents
Novus Biologicals
NBP253197200UG
200 μg Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD44, variant 4
0.2 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page
Unconjugated
Mouse
Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
960
Recombinant fragment corresponding to the v4 domain of human CD44 (exact sequence is proprietary)
Primary
Store at 4C.
CD44v4/1219
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page
Monoclonal
Purified
RUO
Human
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
CD44
IgG2a κ
Protein A or G purified
88 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.