Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Antibody (CD44v4/1700R) - variant 4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP253205100UG
Description
CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD44, variant 4 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
Rabbit | |
88 kDa | |
100 μg | |
Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
960 | |
Store at 4C. | |
IgG κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CD44v4/1700R | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
CD44 | |
Recombinant fragment corresponding to the v4 domain of human CD44 (exact sequence is proprietary) | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction