Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Monoclonal Antibody (IM7), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | CD44 |
---|---|
Clone | IM7 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M010T03B09A
![]() |
Invitrogen™
M010T03B09A |
100 μg |
Each for $375.00
|
|
|||||
M010T02B09A
![]() |
Invitrogen™
M010T02B09A |
25 μg |
Each for $194.00
|
|
|||||
Description
CD44 Monoclonal antibody specifically detects CD44 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
CD44 | |
0.1 mg/mL | |
Monoclonal | |
Rat | |
Human, Mouse | |
P15379, P16070 | |
12505, 960 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
IM7 | |
Flow Cytometry | |
NovaFluor Blue 690 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
IgG2b κ | |
Affinity Chromatography | |
CD44 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title