Learn More
CD44 Mouse anti-Human, Brilliant Violet 421, Clone: 515, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744267
Description
The 515 monoclonal antibody specifically binds to CD44. CD44 is an 80-95 kDa type I transmembrane glycoprotein, also known as Phagocytic glycoprotein-1 (Pgp-1), or Extracellular matrix receptor type III (ECMR-III). CD44 is a member of the hyaladherin family of hyaluronan-binding proteins, with structural similarities to selectins. CD44 is the receptor for hyaluronic acid. CD44 is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is also called and has functional roles in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. The 515 monoclonal antibody can reportedly block cellular adhesion to hyaluronic acid.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
Specifications
CD44 | |
515 | |
Brilliant Violet 421 | |
Pgp-1, CSPG8, ECMR-III, Epican, H-CAM, HCELL, Hermes, HUTCH-1 | |
Human EBV-transformed Arent B Lymphoblastoid Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.