Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Mouse anti-Human, Clone: HI44a (FITC), Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB13922
Description
Mouse monoclonal antibody raised against human CD44.
- High specificity
- High reactivity
Specifications
CD44 | |
Monoclonal | |
FITC | |
Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user. | |
P16070 | |
CD44 | |
Human leukocytes of a patient suffering from LGL type leukemia. | |
100 Reactions | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
HI44a | |
Mouse monoclonal antibody raised against human CD44. | |
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide). | |
CDW44/CSPG8/ECMR-III/HCELL/IN/LHR/MC56/MDU2/MDU3/MGC10468/MIC4/MUTCH-I/Pgp1 | |
Mouse | |
Protein A/G purification | |
RUO | |
960 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction