Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Mouse, anti-Human, R718, Clone: 515, BD OptiBuild™

Catalog No. BDB752028 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752028 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752028 Supplier BD Biosciences Supplier No. 752028
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 515 monoclonal antibody specifically binds to CD44. CD44 is an 80-95 kDa type I transmembrane glycoprotein, also known as Phagocytic glycoprotein-1 (Pgp-1), or Extracellular matrix receptor type III (ECMR-III). CD44 is a member of the hyaladherin family of hyaluronan-binding proteins, with structural similarities to selectins. CD44 is the receptor for hyaluronic acid. CD44 is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 has functional roles in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. The 515 monoclonal antibody can reportedly block cellular adhesion to hyaluronic acid.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone 515
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.