Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Mouse, anti-Human, R718, Clone: L178, BD OptiBuild™

Catalog No. BDB752010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752010 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752010 Supplier BD Biosciences Supplier No. 752010
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L178 monoclonal antibody specifically binds to CD44 which is also known as Homing-associated cell adhesion molecule (H-CAM) or Phagocytic glycoprotein 1 (Pgp-1). CD44 is a single-pass type I transmembrane glycoprotein that exists in a variety of isoforms also known as variant CD44 (CD44v) or CD44R forms. CD44 is synthesized as a 37 kDa polypeptide that is converted to an 80-95 kDa form by glycosylation via N- and O-linkages, or to a 180-200 kDa form by the addition of chondroitin sulfate. It is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. Depending on the cell type, CD44 may participate in a variety of functions including cell adhesion, motility and cell activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone L178
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.