Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Rat anti-Human, Clone: Hermes-1, MilliporeSigma™

Catalog No. MABF209225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2092-25 25 μg
MABF2092-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2092-25 Supplier MilliporeSigma Supplier No. MABF209225UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-CD44, clone Hermes-1 is a rat monoclonal antibody that detects human CD44 and has been tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone Hermes-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CD44,LHR,MDU2,MDU3,MIC4
Host Species Rat
Immunogen Human tonsil lymphocytes.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000601.3
Test Specificity Clone hermes-1 is a rat monoclonal antibody that detects human CD44.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.