Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Std./H-CAM Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS668R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS668R1 1 mL Unconjugated Prediluted Prediluted
MS668P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS668P1 500 μL Unconjugated Concentrated 200μg/mL
MS668P 1 mL Unconjugated Concentrated 200μg/mL
MS668B0 100 μL Biotin Concentrated 200μg/mL
MS668B1 500 μL Biotin Concentrated 200μg/mL
MS668B 1 mL Biotin Concentrated 200μg/mL
MS668PABX 200 μL Unconjugated Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS668R1 Supplier Epredia Supplier No. MS668R1

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunoprecipitation and flow cytometry experiments.

CD44 is a cell surface glycoprotein expressed on lymphocytes, monocytes, and granulocytes. CD44 is also known as homing cell adhesion molecule (H-CAM), Phagocytic glycoprotein-1 (PgP-1), ECM-III, HUTCH-1, or Hermes-1. CD44 has been implicated in cell migration, in lymphocyte homing, and in tumor metastasis. Ab-4 reacts with red cells pretreated with 2-aminoethylisothiouronium bromide (AET). Digestion of intact erythrocytes by trypsin treatment leaves a 50-54 kDa protein fragment attached to the membrane, while chymotrypsin leaves a somewhat similarly sized 51-56 kDa component that are recognized by Ab-4. This antibody enhances IL-2 production by CD2-transfected murine hybridoma stimulated by JY cells while it has no significant effect on IL-2 production by Jurkat cells stimulated with CD3, PMA, or CD2. Ab-4 enhances proliferation of PBL stimulated with CD2.

Host Species: Mouse

Clone: 156-3C11

Isotype: IgG2a

Species Reactivity: Human, Baboon and Green Monkey. Does not react with rat. Others not tested.

Epitope: Resistant to digestion by trypsin and chymotrypsin

Immunogen: Stimulated human leukocytes

Molecular Weight: 80-95 kDa

Positive Control: HUVEC or HeLa cells, tonsil or esophageal CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Flow Cytometry

Specifications

Antigen CD44 Std./H-CAM Ab-4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 156-3C11
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Stimulated human leukocytes
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Non-human Primate
Form Prediluted
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.