Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44, variant 6 Antibody (bivatuzumab) - Humanized, R&D Systems™
SDP

Catalog No. NB465807 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB465807 100 μg
NB465805 50 μg
NB465806 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB465807 Supplier R&D Systems Supplier No. NBP328588100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Human Monoclonal Antibody

CD44, variant 6 Monoclonal specifically detects CD44, variant 6 in Human samples. It is validated for ELISA∣Flow Cytometry∣Functional.

Specifications

Antigen CD44, variant 6
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone bivatuzumab
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P16070
Gene Alias CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Host Species Human
Immunogen CD44v6
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.