Learn More
CD44v10 Monoclonal Antibody (VFF-14)

Description
CD44v10 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | CD44v10 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | VFF-14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CD44 |
| Gene Accession No. | P16070 |
| Gene Alias | AL133330.1; AU023126; AW121933; AW146109; Cd44; CD44 antigen; CD44 molecule (Indian blood group); CD44 splice variant; CD44 var; CD44 variant; CD44A; CD44v10; CDw44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; epican; Extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; H-CAM v10; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCHI; HUTCH-I; Hyaluronate receptor; IN; LHR; Ly-24; Lymphocyte antigen 24; MC56; MDU2; MDU3; METAA; MGC10468; MIC4; MUTCH-I; PGP I; Pgp1; PGP-1; PGP-I; phagocytic glycoprotein 1; Phagocytic glycoprotein I; RHAMM; soluble CD44 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.