Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD44var (v5) Monoclonal Antibody (VFF-8), eBioscience™
GREENER_CHOICE

Catalog No. 5017750
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-177-50 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-177-50 Supplier Invitrogen™ Supplier No. BMS115
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: Specifically recognizes an epitope encoded by exon v5 on the variant portion of human CD44. Standard CD44 (CD44s) lacks the entire variable region and is found mainly in cells of lymphohematopoietic origin. The multiple isoforms (designated CD44v; v = variant) contain alternatively spliced exons inserted within the extracellular domain. These isoforms are known as CD44v1, CD44v2, CD44v3, CD44v4, CD44v5, CD44v6, CD44v7, CD44v8, CD44v9, CD44v10, CD44v11. Applications Tested: Flow Cytometry, Immunohistochemistry (Paraffin), Western Blotting. Purity: >95%.

CD44 cell surface antigen is a 100kDa glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44var (v5)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VFF-8
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CD44
Gene Accession No. P16070
Gene Alias AL133330.1; CD44; CD44 antigen; CD44 molecule (Indian blood group); CD44 splice variant; CD44 var; CD44 variant; CD44v5; CDW44; cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; H-CAM v5; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCHI; HUTCH-I; Hyaluronate receptor; IN; LHR; MC56; MDU2; MDU3; MGC10468; MIC4; MUTCH-I; PGP I; Pgp1; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; soluble CD44
Gene Symbols CD44
Host Species Mouse
Immunogen Rec. Human CD44var (v5).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 960
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.