Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45.1 Antibody (A20), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128070AF405
Description
CD45.1 Monoclonal specifically detects CD45.1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CD45.1 | |
Monoclonal | |
Alexa Fluor 405 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse CD 45.1 (Leukocyte Common Antigen) | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
A20 | |
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
PTPRC | |
SJL mouse thymocytes and splenocytes | |
0.25 mL | |
Innate Immunity, Mesenchymal Stem Cell Markers, Metabotropic Glutamate Receptors, Stem Cell Markers | |
5788 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction