Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD45.1 Monoclonal Antibody (A20), FITC

Mouse Monoclonal Antibody
$125.00 - $650.00
Specifications
| Antigen | CD45.1 |
|---|---|
| Clone | A20 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C, store in dark |
| Applications | Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-113-7633
|
Invitrogen™
A14732 |
25 μg |
Each for $125.00
|
|
|||||
50-113-8109
![]()
|
Invitrogen™
MCD45101 |
500 μg |
Each for $650.00
|
|
|||||
Description
CD45.1 Monoclonal Antibody for Flow
CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.Specifications
| CD45.1 | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| FITC | |
| Mouse | |
| RUO | |
| PBS with <0.1% sodium azide; pH 7.4 | |
| P06800 | |
| 19264 | |
| Mouse CD45.1 from SJL mouse thymocytes and splenocytes. | |
| Antibody |
| A20 | |
| 4°C, store in dark | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Mouse | |
| Ptprc | |
| B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| Ptprc | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title