Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45.1 Mouse anti-Mouse, AF 647, Clone: A20, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 179531
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD45.1 | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
SJL mouse thymocytes and splenocytes | |
RUO | |
Mouse | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
A20 | |
Alexa Fluor 647 | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction