Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD45.2 Monoclonal Antibody (104), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 376045482
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-604-5482 100 μg
37-604-5480 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-604-5482 Supplier Invitrogen™ Supplier No. 376045482
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD45.2 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45.2 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45.2 protein exists as multiple isoforms as a result of alternative splicing; these isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. These isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45.2 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45.2
Applications Flow Cytometry
Classification Monoclonal
Clone 104
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 661
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Ptprc
Gene Accession No. P06800
Gene Alias B220; Cd45; CD45 antigen; CD45R; GP180; LCA; L-CA; leukocyte common antigen; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type, C; Ptprc; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein
Gene Symbols Ptprc
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19264
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.