Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Antibody (30-F11), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10077417C
Description
CD45 Monoclonal specifically detects CD45 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Product Image.Specifications
CD45 | |
Monoclonal | |
0.5mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
PTPRC | |
Mouse thymus or spleen | |
0.1 mL | |
Adaptive Immunity, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
5788 | |
Human, Mouse, Rat | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
30-F11 | |
DyLight 650 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Recognizes all CD45 isoforms as well as the known mouse CD45 alloantigens (CD45-1/Ly 5a and CD45-2/Ly 5b) | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction