Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Antibody (IBL-3/16), Novus Biologicals™

Rat Monoclonal Antibody
$207.00 - $499.50
Specifications
| Antigen | CD45 |
|---|---|
| Clone | IBL-3/16 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50 - 1:100 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017713
![]() |
Novus Biologicals
NB100653640.025MG |
0.025 mg |
Each for $207.00
|
|
|||||
NB10065364
![]() |
Novus Biologicals
NB10065364 |
0.125 mg |
Each for $499.50
|
|
|||||
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| CD45 | |
| 0.5 mg/mL | |
| Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| P06800 | |
| 5788 | |
| Purified B cells from mouse lymph nodes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| IBL-3/16 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50 - 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG1 | |
| Protein G purified | |
| Recognizes mouse CD45. The enhances the MHC Class II induced cluster formation of mouse B cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title