Learn More
CD45 Antibody (LT40), PE, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP128298
Catalog No. NBP128298
Only null left
Add to Cart
Description
Description
CD45 Monoclonal specifically detects CD45 in Chicken samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications
Specifications
CD45
Monoclonal
0.1 mg/mL
Flow Cytometry <= 0.2 ug/10^6 cells, Immunoprecipitation 1:10-1:500
PTPRC
Chicken bursal lymphocytes
0.1 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
5788
Chicken
Purified
Flow Cytometry, Immunoprecipitation
LT40
PE
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgM purified
RUO
Primary
Chicken CD45 (Mr 190 to 215-kDa).
Store at 4C. Do not freeze.
IgM κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only