Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Antibody (rPTPRC/1147), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32036420UG
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
CD45 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen | |
Recombinant full-length human CD45 protein | |
20 μg | |
Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
Western Blot, Immunohistochemistry (Paraffin) | |
rPTPRC/1147 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction