| Antigen |
CD45 |
| Applications |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification |
Monoclonal |
| Clone |
SPM496 |
| Concentration |
0.2mg/mL |
| Conjugate |
Unconjugated |
| Dilution |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Formulation |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide |
| Gene Symbols |
PTPRC |
| Host Species |
Mouse |
| Immunogen |
Stimulated human leukocytes |
| Purification Method |
Protein A or G purified |
| Quantity |
0.02 mg |
| Regulatory Status |
RUO |
| Research Discipline |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
5788 |
| Test Specificity |
CD45R, also designated CD45 and PTPRC, has been identified as a transmembrane glycoprotein, broadly expressed among hematopoietic cells. Multiple isoforms of CD45R are distributed throughout the immune system according to cell type. These isoforms arise because of alternative splicing of exons 4, 5, and 6. The corresponding protein domains are characterized by the binding of monoclonal antibodies specific for CD45RA (exon 4), CD45RB (exon 5), CD45RC (exon 6) and CD45RO (exons 4 to 6 spliced out). The variation in these isoforms is localized to the extracellular domain of CD45R, while the intracellular domain is conserved. CD45R functions as a phosphor-tyrosine phosphatase. This MAb reacts with all isoforms of CD45R expressed by all hematopoietic cells, except erythrocytes, having a higher level of expression on lymphocytes than on granulocytes (Workshop IV). Epitope of this MAb resides in the sialic acid moiety of T200 molecule. It enhances the proliferative response of B cells in an anti-immunoglobulin-induced B cell proliferation. Antibody to CD45 is useful in differential diagnosis of lymphoid tumors from non-hematopoietic undifferentiated neoplasms. |
| Target Species |
Human |
| Content And Storage |
Store at 4C. |
| Form |
Purified |
| Isotype |
IgG1 κ |
|
Show More
Show Less
|
|