Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Monoclonal Antibody (2D1), NovaFluor™ Yellow 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$209.00 - $463.00
Specifications
Antigen | CD45 |
---|---|
Clone | 2D1 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H005T03Y03A
![]() |
Invitrogen™
H005T03Y03A |
100 Tests |
Each for $463.00
|
|
|||||
H005T02Y03A
![]() |
Invitrogen™
H005T02Y03A |
25 Tests |
Each for $209.00
|
|
|||||
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow CytometrySpecifications
CD45 | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P08575 | |
5788 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
2D1 | |
Flow Cytometry | |
NovaFluor Yellow 610 | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen; T220 and B220 | |
IgG1 κ | |
Affinity Chromatography | |
PTPRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title