Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Monoclonal Antibody (4E9B2), CoraLite488, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48860287100UL
Description
CD45 Monoclonal antibody specifically detects CD45 in Human, Mouse, Pig samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)Specifications
| CD45 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| B220; CD45; CD45R; GP180; L-CA; LCA; loc; Ly-5; LY5; Lyt-4; PTPRC; T200 | |
| Protein A | |
| RUO | |
| 5788 | |
| -20° C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 4E9B2 | |
| CoraLite 488 | |
| P08575 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Pig | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction