Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Monoclonal Antibody (CD45-2B11), eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

$144.00 - $1562.00

Specifications

Antigen CD45
Clone CD45-2B11
Concentration 0.5 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-255-0879
Encompass_Preferred
View Documents
Invitrogen™
14945737
2 mg
Each for $1,562.00
Only null left
Add to Cart
 
50-112-2784
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
14945782
100 μg
Each for $144.00
Only null left
Add to Cart
 
Description

Description

The CD45-2B11 MAb reacts with human CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding circulating erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in lymphocyte proliferation and differentiation. The 2B11 antibody is useful for recognition of normal and neoplastic lymphoid cells. Applications Reported: This CD45-2B11 antibody has been reported for use in flow cytometric analysis, immunoblotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This CD45-2B11 antibody has been tested by immunohistochemisty on formalin-fixed paraffin embedded tissue using high pH antigen retrieval. This can be used at less than or equal to 10 μg/mL. This CD45-2B11 antibody has also been tested by western blot at 5 μg/mL and by flow cytometric analysis on human peripheral blood cells at 0.25 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.
Specifications

Specifications

CD45
0.5 mg/mL
Unconjugated
Mouse
Human
5788
Primary
Antibody
CD45-2B11
Monoclonal
Liquid
RUO
P08575
IgG1 κ
Affinity chromatography
PTPRC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.