Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Mouse anti-Human, BB515, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564585
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
Specifications
CD45 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
HI30 | |
Vol. Per test: 5μL | |
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5 | |
Human Peripheral Blood Leucocytes | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction