Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Human, BUV805, Clone: HI30, BD Horizon™

Mouse Monoclonal Antibody

$248.00 - $470.00

Specifications

Antigen CD45
Clone HI30
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 805
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612892
View Documents
BD Biosciences
612892
25 Tests
Each for $248.00
Only null left
Add to Cart
 
BDB612891
View Documents
BD Biosciences
612891
100 Tests
Each for $470.00
Only null left
Add to Cart
 
Description

Description

CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD45
Flow Cytometry
Brilliant Ultraviolet 805
RUO
IgG1 κ
Refrigerate (2 to 8°C)
HI30
Monoclonal
Mouse
Human
Primary
CD45
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.