Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Mouse anti-Human, Horse, APC, Clone: F10-89-4, Southern Biotech™

Mouse Monoclonal Antibody
$144.20 - $313.50
Specifications
Antigen | CD45 |
---|---|
Clone | F10-89-4 |
Concentration | Lot specific |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD45 | |
Lot specific | |
Monoclonal | |
Mouse | |
Equine, Human | |
Purified T cells from human lymph nodes | |
Primary |
F10-89-4 | |
Flow Cytometry | |
APC | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG2a | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title