Learn More
CD45 Mouse, anti-Human, R718, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566961
Description
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

Specifications
CD45 | |
Monoclonal | |
R718 | |
100 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
HI30 | |
Mouse | |
RUO | |
Human | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.