Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Mouse anti-Human, RB545, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
$162.00 - $398.00
Specifications
Antigen | CD45 |
---|---|
Clone | HI30 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB545 |
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometrySpecifications
CD45 | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human | |
PTPRC; LCA; L-CA; Leukocyte Common Antigen; T200; GP180; LY5 | |
Human Peripheral Blood Leucocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
HI30 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
5788 | |
IgG1 κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title