Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Rat anti-Mouse, BB700, Clone: 30-F11, BD Horizon™
Rat Monoclonal Antibody
$124.50 - $299.50
Specifications
Antigen | CD45 |
---|---|
Clone | 30-F11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD45 | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
30-F11 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Rat | |
Mouse | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title