Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Rat anti-Mouse, BV 786, Clone: I3/2.3 (also known as I3/2 or I3), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 752409
This item is not returnable.
View return policy
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry
Specifications
CD45 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2b λ |
Flow Cytometry | |
I3/2.3 (also known as I3/2 or I3) | |
Brilliant Violet 786 | |
Cd45; L-CA; LCA; Ly-5; Ly5; Ptprc | |
Mouse Lymphoma Cell Line | |
RUO | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction