Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45, Rat Anti-Mouse, RY610, Clone: 30-F11, BD Horizon™

Catalog No. BDB571166 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg;
25 μg;
Conjugate:
RY610
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571166 25 μg; RY610
BDB571165 100 μg; RY610
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571166 Supplier BD Biosciences Supplier No. 571166
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45 Monoclonal antibody specifically detects CD45 in Mouse (QC Testing) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone 30-F11
Concentration 0.2 mg/mL
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ptprc; LCA; Leukocyte common antigen; T200; Ly-5; Lyt-4
Host Species Rat
Immunogen Mouse Thymus / Spleen
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 25 μg;
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19264
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.