Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45R (B220) Rat anti-Mouse, Human, BUV737, Clone: RA3-6B2, BD Horizon™

Catalog No. BDB612839 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612839 25 μg
BDB612838 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612839 Supplier BD Biosciences Supplier No. 612839
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD45R (B220) Monoclonal antibody specifically detects CD45R (B220) in Mouse,Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45R (B220)
Applications Flow Cytometry
Classification Monoclonal
Clone RA3-6B2
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Host Species Rat
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD45R (B220)
Target Species Mouse, Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.