Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Antibody (PTPRC/4367R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32037720UG
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD45RA | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Recombinant fragment (around aa1-600) of human CD45RA protein (extracellular domain) | |
20 μg | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
PTPRC/4367R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction