Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD45RA Monoclonal Antibody (HI100), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™

Catalog No. H014T02B08A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H014T02B08A 25 Tests
H014T03B08A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H014T02B08A Supplier Invitrogen™ Supplier No. H014T02B08A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing and these isoforms differ in their extracellular domains, however, they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage, with the exception of mature red cells and is not detected on differentiated cells of other tissues. It is likely that CD45 plays an important role in signal transduction, inhibition or upregulation of various immunological functions. Diseases associated with CD45 dysfunction include severe combined immunodeficiency and Hepatitis C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry
Classification Monoclonal
Clone HI100
Concentration 0.8 μg/Test
Conjugate NovaFluor Blue 660-120S
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene PTPRC
Gene Accession No. P08575
Gene Alias B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; Leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; PTPRC; Receptor-type tyrosine-protein phosphatase C; RT7; T100; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Symbols PTPRC
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.