Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Mouse anti-Human, BUV395, Clone: 5H9, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569489
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow cytometry
Specifications
CD45RA | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
5H9 | |
Brilliant Ultraviolet 395 | |
CD45R; PTPRC; LCA; Leukocyte common antigen | |
Affinity Chromatography | |
RUO | |
5788 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction