Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Mouse anti-Human, BUV563, Clone: HI100, BD Horizon™
Mouse Monoclonal Antibody
$249.00 - $472.00
Specifications
Antigen | CD45RA |
---|---|
Clone | HI100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry.Specifications
CD45RA | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
HI100 | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD45RA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title