Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Mouse anti-Human, Clone: 4KB5, Abnova™

Catalog No. 89933827 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-827 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-827 Supplier Abnova Corporation Supplier No. MAB12721

Mouse monoclonal antibody raised against human CD45RA.

Specifications

Antigen CD45RA
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4KB5
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Host Species Mouse
Immunogen Hairy cell leukemia cells
Purification Method Protein G purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.