Learn More
CD45RA Mouse anti-Human, PE-CF594, Clone: HI100, BD
Mouse Monoclonal Antibody
$198.00 - $422.00
Specifications
Antigen | CD45RA |
---|---|
Clone | HI100 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-CF594 |
Description
The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Specifications
CD45RA | |
Flow Cytometry | |
PE-CF594 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HI100 | |
Monoclonal | |
Mouse | |
Human | |
IgG2b κ | |
Affinity Purified | |
CD45R; PTPRC; LCA; Leukocyte common antigen |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.