Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Mouse, anti-Human, R718, Clone: 5H9, BD OptiBuild™

Catalog No. BDB752257 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752257 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752257 Supplier BD Biosciences Supplier No. 752257
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5H9 monoclonal antibody specifically binds to the human form of the leukocyte common antigen (LCA), CD45RA. This clone also crossreacts with CD45RA expressed by peripheral blood lymphocytes, monocytes and some granulocytes from rhesus macaque, baboon, and cynomolgus monkeys. The observed frequency of CD45RA positive lymphocytes is higher in the non-human primate compared to what is seen with normal human donor lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry
Classification Monoclonal
Clone 5H9
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.